Learn More
Idex Low-Pressure PEEK Tees
Combine, split, or intersect fluid streams throughout your system
Supplier:  Idex UP P 712/DUP
Encompass
 View more versions of this product
Catalog No. 05700177
Only null left
Description
Description
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
Use Tee connectors in a variety of low-pressure situations throughout your fluidic system. Handy mounting holes make it a snap to mount connectors. Tee connector assemblies come complete with nuts and ferrules.
Specifications
Chemical Identifiers
1000 psi
PEEK
Specifications
Tees and Crosses
1000 psi
0.50 mm
0.020 in.
PEEK
Product Suggestions
Product Suggestions