missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Low Form Cylindrical Dewar Flask with Aluminum Housing

Curved bottom helps to increase coolant capacity

$188.82 - $1,440.64

Specifications

Color Silver
Material Aluminum
For Use With (Application) Liquid Nitrogen
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
FB1019621
View Documents
Fisherbrand™
FB1019621
150 mL N/A
Only null left
 
FB1019620
View Documents
Fisherbrand™
FB1019620
220 mL N/A
Only null left
 
FB101966
View Documents
Fisherbrand™
FB101966
350 mL N/A
Only null left
 
FB101967
View Documents
Fisherbrand™
FB101967
850 mL N/A
Only null left
 
FB101968
View Documents
Fisherbrand™
FB101968
2500 mL N/A
Only null left
 
Description

Description

  • Cylindrical
  • Low Form
  • Aluminum Housing
  • Borosilicate Glass with Silvered Double Walls
Specifications

Specifications

Silver
Aluminum
Liquid Nitrogen
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'