Learn More
Boca Scientific Inc LowCross-Buffer, 500ml - Minimize nonspecific binding, cross-reactivities and matrix effects in immunoassays.
Catalog No. NC9048027
Encompass
LowCross-Buffer is an innovative assay buffer for ELISA, EIA, RIA, protein arrays, Western blotting, immunohistochemistry and immuno-PCR. It is ready-to-use and can replace your standard sample or antibody dilution buffer. The formulation of LowCross-Buffer helps to reduce immunoassay interference and can therefore greatly improve the quality of detection. Nonspecific binding of the antibodies, negative effects of interfering substances, and low or medium affinity cross reactivities of the antibodies can be significantly decreased. Matrix effects, such as those from blood sera or plasma specimens, are minimized. Reduction of these negative effects improves the quality of the assay and the reliability of the results.
Catalog No. NC9048027 Supplier Boca Scientific Inc Supplier No. 100 500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
LowCross-Buffer is an innovative assay buffer for ELISA, EIA, RIA, protein arrays, Western blotting, immunohistochemistry and immuno-PCR. It is ready-to-use and can replace your standard sample or antibody dilution buffer. The formulation of LowCross-Buffer helps to reduce immunoassay interference and can therefore greatly improve the quality of detection. Nonspecific binding of the antibodies, negative effects of interfering substances, and low or medium affinity cross reactivities of the antibodies can be significantly decreased. Matrix effects, such as those from blood sera or plasma specimens, are minimized. Reduction of these negative effects improves the quality of the assay and the reliability of the results.