missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Solution: PAN; 0.1%
Pre-measured, unit dose reagent for water quaility testing
Supplier: Lovibond™ 530631
Description
- Specifically formulated to serve as direct replacement to Hach™ PAN Indicator Solution
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
| Dark Orange | |
| 100 mL | |
| Water quality testing | |
| 100 mL | |
| 3.38 oz. |
| 8 | |
| Fluid | |
| MC500, MD600 colorimeter, SP600 spectrophotometer, SC450, MD100 Manganese colorimeters and Manganese low range reagent set | |
| PAN Indicator Solution |