Learn More
Lovibond™ Solution: Alkaline-Cyanide
Pre-measured, unit dose reagent for water quaility testing
$88.17 - $131.31
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|
09-053-207
|
Lovibond™
530620 |
50 mL |
Each for $88.17
|
|
|||||
|
09-534-454
|
Lovibond™
530621 |
100 mL |
Each for $131.31
|
|
|||||
Description
- Specifically formulated to serve as direct replacement to Hach™ Alkaline Cyanide Reagent Solution
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
| Alkaline Cyanide Solution |