missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Phosphate No. 2 LR Tablets

Catalog No. 09053192
Change view
Click to view available options
Range:
0.05 to 4mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09053192 0.05 to 4mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09053192 Supplier Lovibond™ Supplier No. 513050BT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lovibond Replacement Reagents for CHECKIT Comparator: Phosphate No. 2 LR Tablets, Range: 0.05 to 4mg/L, White, Composition: Boric Acid 70 to 80%, Disodium Disulphite 10 to 20%, Ammonium Heptamolybdate Tetrahydrate 5 to 10%, non-hazardous components 2.5%, Format: Tablet

  • The Phosphate No. 2 LR tablet reagent is an indicator system designed for precise measurement in pool test kits.
  • Easy Handling: Tablet form allows for safe and simple handling.
  • Precise Dosing: Facilitates accurate and consistent dosing.
  • Durability: Provides long shelf life and enormous durability.
  • Packaging: Provided in an aluminium blister pack.
  • Method: M2538

Specifications

Color White
Composition Boric Acid 70 to 80%, Disodium Disulphite 10 to 20%, Ammonium Heptamolybdate Tetrahydrate 5 to 10%, non-hazardous components 2.5%
pH Range 4.72
Format Tablet
For Use With (Application) Low range phosphate AC test kit, phosphate, ortho LR reagent set, water analysis
For Use With (Equipment) MC500 colorimeters and SP600 spectrophotometers
Range 0.05 to 4mg/L
Test Method Phosphomolybdic Acid/Ascorbic Acid
Type Phosphate No. 2 LR Tablet