Learn More
Lovibond™ Reagent Set: Powder Pack; Oxygen Scavenger; DEHA
Premeasured unit dose reagent for water quality testing
Supplier: Lovibond™ 536000
Includes: Oxyscav Reagent (10mL sample, 100 pack), DEHA 2 Reagent Solution (100mL)
Description
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Orbeco-Hellige and Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
| Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
| 10mL Oxyscav reagent (100 pack), 10mL DEHA 2 reagent solution | |
| 20 to 500μg | |
| DEHA Reagent Set |
| Hach™ and other manufacturers test kits, colorimeters and spectrophotometers | |
| 100 | |
| PPST |