Learn More
Lovibond™ Ammonia Reagent Set Powder Packs™
Description
Includes
Ammonia Cyanurate Reagent (10mL sample, 100 pack), Ammonia Salicylate Reagent (10mL sample, 100 pack)
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | Hach™ and other manufacturers test kits, colorimeters and spectrophotometers |
| Includes | 10mL ammonia cyanurate reagent (100 pack), 10mL ammonia salicylate reagent (100 pack) |
| No. of Tests | 100 |
| Range | 0 to 0.8mg/L |
| Test Method | Salicylate |
| Type | Ammonia Nitrogen Reagent Set |