missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Process Chlorine Analyzer Reagents: DPD Indicator Powder
Description
- Used for 30 day continuous supply
Specifications
Specifications
| Format | Powder |
| For Use With (Equipment) | Hach™ CL17™ process chlorine analyzer |
| Quantity | 24 g |
| Type | DPD Indicator Powder |