missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Total Chlorine Powder Packs™
Premeasured unit dose DPD reagents for water quality testing
$91.00 - $871.37
Specifications
| Certifications/Compliance | US EPA |
|---|---|
| Format | Powder |
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer |
| Type | DPD Total Chlorine Reagent |
| Catalog Number | Mfr. No. | Quantity | Range | Volume (English) | Volume (Metric) | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Range | Volume (English) | Volume (Metric) | Price | Quantity | ||
|
09-053-214
|
Lovibond™
530080 |
100 Pack | 0 to 3.5 mg/L | 0.16 oz. | 5 mL |
Each for $91.00
|
|
||
|
09-053-223
|
Lovibond™
530123 |
1000 Pack | 0 to 2 mg/L | 0.33 oz. | 10 mL |
Pack of 1000 Each for $752.60
|
|
||
|
09-053-222
|
Lovibond™
530120 |
100 Pack | 0 to 2 mg/L | 0.33 oz. | 10 mL |
Each for $97.97
|
|
||
|
09053224
|
Lovibond™
530130 |
100 Pack | 0 to 5 mg/L | 0.84 oz. | 25 mL |
Each for $100.74
|
|
||
|
09-053-225
|
Lovibond™
530133 |
1000 Pack | 0 to 5 mg/L | 0.84 oz. | 25 mL |
Each for $871.37
|
|
||
Description
Premeasured unit dose DPD reagents for water quality testing
- For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
- Also suitable for use in Hach™ instruments and test kits
- Easy-to-open, sealed foil packets eliminate risk of cross-contamination
- Available for 5, 10, and 25mL sample sizes
- Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
| US EPA | |
| Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
| DPD Total Chlorine Reagent |
| Powder | |
| SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer |