Learn More
Lovibond™ Powder Packs™: Silica HR Acid
Pre-measured, unit dose reagents for water quaility testing
Supplier:  Lovibond™ 531703
Catalog No. 09-053-750
Only null left
Explore available promotions
Description
Description
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open the package
  • Easily create a “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in the corners
Specifications
Specifications
Water quality testing
Colorimeter
0 to 100mg/L
HR Acid Reagent
Product Suggestions
Product Suggestions