missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Powder Packs™ for Free Chlorine
Premeasured unit dose DPD reagents for water quality testing
$89.09 - $875.82
Specifications
| Certifications/Compliance | US EPA |
|---|---|
| Format | Powder |
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer |
| Type | DPD Free Chlorine Reagent |
| Catalog Number | Mfr. No. | Quantity | Range | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Range | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||
|
09053216
|
Lovibond™
530090 |
100 Pack | 0 to 3.5 mg/L | 0.16 oz. | 5 mL |
Each for $89.09
|
|
||
|
09053219
|
Lovibond™
530103 |
1000 Pack | 0 to 2 mg/L | 0.33 oz. | 10 mL |
Each for $821.97
|
|
||
|
09053218
|
Lovibond™
530100 |
100 Pack | 0 to 2 mg/L | 0.33 oz. | 10 mL |
Each for $99.74
|
|
||
|
09053217
|
Lovibond™
530093 |
1000 Pack | 0 to 3.5 mg/L | 0.16 oz. | 5 mL |
Each for $712.37
|
|
||
|
09053220
|
Lovibond™
ORB530110 |
100 Pack | 0 to 5 mg/L | 0.84 oz. | 25 mL |
Each for $106.40
|
|
||
|
09053221
|
Lovibond™
530113 |
1000 Pack | 0 to 5 mg/L | 0.84 oz. | 25 mL |
Each for $875.82
|
|
||
Description
Premeasured unit dose DPD reagents for water quality testing
- For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
- Also suitable for use in Hach™ instruments and test kits
- Easy-to-open, sealed foil packets eliminate risk of cross-contamination
- Available for 5, 10, and 25mL sample sizes
- Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
| US EPA | |
| Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
| DPD Free Chlorine Reagent |
| Powder | |
| SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer |
Safety and Handling
ShelfLife : 5 Years