missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD 100 Cooling Water Colorimeter

Catalog No. 09053676
Change view
Click to view available options
Resolution:
0.01 A
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Resolution
09053676 0.01 A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09053676 Supplier Lovibond™ Supplier No. 276240

Includes

Instrument in carrying case, 4 x AA batteries, three round glass vials (24mm) with lids, crushing/stirring rod, Guarantee sheet, Certificate (COC), Instruction manual

One colorimeter tests multiple parameters associated with cooling water.

One photometer tests multiple parameters associated with cooling water.

  • Ideal for routine, on-site system monitoring
  • Photometric measurement for accurate, repeatable results
  • Easy-to-use, push button technology with step by step guides
  • Negates the need for up to 13 seperate pieces of equipment
  • Select range of required reagents to complete your kit
  • Combine with pH, conductivity and drop test chemistries - complete analysis program

Specifications

  • Optics: LEDs, interference filters (IF) and photo sensor in transparent sample chamber.
  • Wavelength Accuracy: ±1nm
  • Photometric Accuracy: 3% FS (T = 2° to 25°C)
  • Power Supply: 4 micro batteries (AAA), capacity approx. 17 hours or 5000 tests
  • Auto - OFF: automatic switch-off
  • Display: Backlit LCD (on keypress)
  • Storage: internal ring memory for 16 data sets
  • Interfaces: Infrared interface for test data transfer
  • Additional feature: real time clock and date
  • Calibration: factory calibration and user calibration
  • Dimensions (L x W x H): 155 x 75 x 35mm
  • Weight: basic unit approx. 260g

Certifications

CE marked

Specifications

Accuracy 3% Full scale
Calibration Yes
Certifications/Compliance CE
Data Storage Internal ring memory for 16 data sets
Display Backlit LCD
Dimensions (L x W x H) 155 x 75 x 35 mm
Light Source LED
For Use With (Application) Testing multiple parameters associated with cooling water
Height (English) 1.3 in.
Height (Metric) 35 mm
Includes Instrument in carrying case, 4 x AA batteries, three round, 24mm dia. glass vials with lids, crushing/stirring rod, Guarantee sheet, Certificate (COC), Instruction manual
Length (English) 6.1 in.
Length (Metric) 155 mm
Resolution 0.01 A
Temperature Range (Metric) 5°C to 40°C
Weight (English) 0.5 lb.
Weight (Metric) 260 g
Width (English) 3 in.
Width (Metric) 75 mm
Show More Show Less