Learn More
Lovibond™ Legionella Industrial Test Kit
The Legionella System Test Kit allows you to easily collect a filtered water sample directly from the water system. The filtered sample increases the sensitivity of the test.
Supplier: Lovibond™ 56B006101
Description
This Legionella Test Kit contains a sample point fitting to connect to the water system (such as a shower head or on-line sample point) and the system’s pressure drives the sample filtration. Pre-filled syringes contain the required recovery buffer, so there is no need to measure or dilute solutions.
With results in 30 to 35 minutes (including sample prep-time), the test allows for immediate understanding and response to the water system conditions. Regular use will increase confidence in your Legionella control procedures and reduce risk of outbreaks and litigation.
- Ideal for online or industrial water systems
- Collect sample directly from water system
- Allows rapid response to the water conditions
Specifications
| Detecting legionella in a water sample | |
| 5 | |
| Legionella System Test Kit |
| 5 test strips with dispensing pipettes, 5 syringes with recovery buffer, sample collection jar, 5 single use filters, female threaded sample point fitting, and instructions in a carrying case | |
| Lateral flow immunochromatographic assay |