Learn More
Lovibond™ Legionella Field Test Kit
Description
The Legionella Field Test Kit is is easy and fast — no reagents or interpretation of results required! Simply collect a water sample in the sterile bag and dispense onto the test strip. With results in 25 minutes, you'll have confidence that treated water in the distribution network meets standards and does not pose a public health risk.
Legionella can proliferate in water distribution networks and in building plumbing systems even after it has been properly treated at a water treatment plant. Biofilms that form along the distribution network provide an ideal breeding ground for Legionella bacteria.
- Fast, easy and accurate – results in 25 minutes with no sample prep
- Convenient for field testing – no need to send samples to the lab and wait for results
- Allows rapid response to the water conditions
- Regular use will increase confidence in your Legionella control procedures
Specifications
Specifications
| For Use With (Application) | Immediate understanding and response to the water conditions |
| Includes | 10 test strips, 10 dispensing pipettes, 10 sterile sample bags, instruction manual |
| No. of Tests | 10 |
| Test Method | Lateral flow immunochromatographic assay |
| Type | Legionella Field Test Kit |