missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lotus Tetragonolobus (Asparagus Pea) Lectin (LTL), fluorescein (FITC)

Catalog No. L32480
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L32480 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L32480 Supplier Invitrogen™ Supplier No. L32480

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bright-green LTL-fluorophor conjugate

  • Lotus Tetragonolobus Lectin (LTL), fluorescein (FITC), is a bright-green LTL-fluorophor conjugate. LTL is isolated from Tetragonolobus purpureus, also known as Lotus tetragonolobus, within the common names asparagus pea or winged pea.
  • Combined with other lectins, it has been used for the detection of certain diseased tissue in kidney and the detection of colon cancer. LTL has also been used in studies of the inhibition of macrophage migration and in comparison to macrophage inhibition factor (MIF).
  • Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
  • Applications: immunofluorescence, glycobiology
  • Sugar specificity: fucose, arabinose
  • Inhibiting/eluting sugar: 50–100 mM L-fucose
  • Blood group specificity: non-specific, O<A2
  • Unconjugated lectin molecular weight: 107 KDa
  • LTL-FITC is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide. 0.1 mM Ca2+
  • A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.

Specifications

Quantity 2 mg
Source Asparagus pea
Conjugate FITC
Content And Storage When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after reciept.
Label Type Classic Dyes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Subtype Lectins

For Research Use Only. Not for use in diagnostic procedures.