missing translation for 'onlineSavingsMsg'
Learn More

Lornoxicam 98.0+%, TCI America™

Catalog No. p-7131887
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02391G 1 g
L02395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02391G Supplier TCI America Supplier No. L02391G
Only null left

Chemical Identifiers

CAS 70374-39-9
Molecular Formula C13H10ClN3O4S2
Molecular Weight (g/mol) 371.81
MDL Number MFCD00866163
InChI Key WLHQHAUOOXYABV-UHFFFAOYSA-N
Synonym 6-Chloro-4-hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-Dioxide
PubChem CID 54690031
ChEBI CHEBI:31783
IUPAC Name 6-chloro-4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-ylthieno[2,3-e]thiazine-3-carboxamide
SMILES CN1C(=C(C2=C(S1(=O)=O)C=C(S2)Cl)O)C(=O)NC3=CC=CC=N3

Specifications

Melting Point 230°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 371.81
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Lornoxicam
RTECSNumber XJ9095000
TSCA No
Recommended Storage Freezer (-20 °C)