Learn More
SeaPlaque™ Agarose, Lonza™
Description
- For preparative DNA and RNA electrophoresis
- Molecular biology grade
- ≤0.10 EEO
- Low melting temperature argarose (26°–30°C Gelling temperature)
- DNase/RNase-free
Specifications
Specifications
| Chemical Name or Material | SeaPlaque™ Agarose |
| Quantity | 25 g |