missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ RNA Markers
Description
- Consist of RNA transcripts in sizes of 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9kb
- Can be denatured with standard procedures
- Can be visualized on Northern blots with labeled lambda sequence
- Can be detected with ethidium bromide or nucleic acid stains
- Store at -20°C for up to six months or at -80°C for up to 24 months
Specifications
Specifications
| Concentration | 1 μg/mL |
| For Use With (Application) | Size determination of RNA fragments |
| No. of Bands | 10 |
| Quantity | 50 μg |
| Storage Requirements | −20°C |
| Product Type | RNA Molecular Weight Marker |