Learn More
Lonza™ FlashGel™ System Components and Packs: FlashGel Dock
Description
- Provides nearly instant results
- Load samples, watch bands migrate and get data in as little as two minutes
- Includes DNA recovery and gel documentation
- Needs no gel preparation, band excision, purification, and UV light
- Complete separation, recovery and documentation safely at the bench
- Highly sensitive system allows a 5 to 20X reduction in DNA levels, saving time and money
- Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
- Detect <0.1ng DNA or <10ng total RNA
- Clean, sharp separation and straight, uniform sample lanes
FlashGel Dock
- For use with all FlashGel cassettetypes
- Provides electrophoresis and visualization of both DNA and RNA cassettes
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
Specifications
| Type | FlashGel Dock |
| Includes | FlashGel Dock |