Learn More
Longwave UV Mini Light Source
Catalog No. S99188
Change view
Click to view available options
Product Type:
Replacement Bulb
Catalog No. Product Type
S99188 Replacement Bulb
1 options
Catalog No. S99188 Supplier Customer Channels Group (Ccg) Supplier No. 6762
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Requires
Four AA alkaline batteries (S43921, not included)
For use in crime scene search and forensic applications.
  • Ideal for theft detection in viewing invisible stains, powders and pastes
  • Fully portable, battery-operated longwave UV source housed in rugged ABS plastic
  • Peak wavelength: 365nm
Order Info
Dimensions: 6.25H x 2W x 0.75 in. D (15.9 x 5.1 x 1.9cm). Replacement bulb available separately.
Specifications
For Use With (Application) For uv longwave mini light source
Requires Four AA alkaline batteries
Product Type Replacement Bulb