Learn More
New England Biolab Ltd LongAmp Taq 2X Master Mix 500 reactions (50 l vol)
Catalog No. 50995180
Encompass
The LongAmp Taq 2X Master Mix combines high quality NEB recombinant Taq DNA Polymerase with a trace amount of Deep Vent DNA Polymerase. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. This ready-to-use mix offers two fold higher fidelity than Taq DNA Polymerase alone. A wide range of PCR products can be generated, up to 30 kb from lambda or human genomic DNA.
Catalog No. 50995180 Supplier New England Biolab Ltd Supplier No. M0287L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The LongAmp Taq 2X Master Mix combines high quality NEB recombinant Taq DNA Polymerase with a trace amount of Deep Vent DNA Polymerase. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. This ready-to-use mix offers two fold higher fidelity than Taq DNA Polymerase alone. A wide range of PCR products can be generated, up to 30 kb from lambda or human genomic DNA.