Learn More
Dynalon™ Long Spout Graduated Dropping Bottles
Low-density polyethylene bottles with closure and spout.
$143.56 - $143.56
Specifications
Includes Long spout
Material LDPE
Certifications/Compliance DIN 13316 and 168
Graduated Yes
Shape Round
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S01985
View Documents
Dynalon™
22412500
50 mL
Pack of 10 Each for $143.56
Only null left
Description
Description
  • Graduated
  • Can be secured using shoulder and cap tag to avoid sample contamination in transit
  • Supplied with LDPE cap for tip closure
  • An ideal bottle for water and oils as there are no shoulders to restrict the flow
Compliance
Conforms to DlN 13316 and 168.
Specifications
Specifications
Long spout
DIN 13316 and 168
Round
LDPE
Yes
Screw Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'