missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lomefloxacin Hydrochloride 98.0+%, TCI America™
$177.08 - $479.81
Chemical Identifiers
| CAS | 98079-52-8 |
|---|---|
| Molecular Formula | C17H20ClF2N3O3 |
| Molecular Weight (g/mol) | 387.812 |
| MDL Number | MFCD00214312 |
| InChI Key | KXEBLAPZMOQCKO-UHFFFAOYSA-N |
| Synonym | 1-Ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid Hydrochloride |
| PubChem CID | 68624 |
| ChEBI | CHEBI:6518 |
| IUPAC Name | 1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride |
| SMILES | CCN1C=C(C(=O)C2=CC(=C(C(=C21)F)N3CCNC(C3)C)F)C(=O)O.Cl |
Chemical Identifiers
| 98079-52-8 | |
| 387.812 | |
| KXEBLAPZMOQCKO-UHFFFAOYSA-N | |
| 68624 | |
| 1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride |
| C17H20ClF2N3O3 | |
| MFCD00214312 | |
| 1-Ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid Hydrochloride | |
| CHEBI:6518 | |
| CCN1C=C(C(=O)C2=CC(=C(C(=C21)F)N3CCNC(C3)C)F)C(=O)O.Cl |
Specifications
| 98079-52-8 | |
| C17H20ClF2N3O3 | |
| 5 g | |
| KXEBLAPZMOQCKO-UHFFFAOYSA-N | |
| 1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride | |
| 68624 | |
| 387.81 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00214312 | |
| 1-Ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid Hydrochloride | |
| CCN1C=C(C(=O)C2=CC(=C(C(=C21)F)N3CCNC(C3)C)F)C(=O)O.Cl | |
| 387.812 | |
| CHEBI:6518 | |
| ≥98.0% (HPLC,T) | |
| Lomefloxacin Hydrochloride |
Safety and Handling
RTECSNumber : VB1997500
TSCA : No