missing translation for 'onlineSavingsMsg'
Learn More

Lofexidine hydrochloride, 98+%

Catalog No. AAJ6396003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6396003 1 g
AAJ6396006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6396003 Supplier Thermo Scientific Chemicals Supplier No. J6396003

Alpha-2 adrenergic agonist | CAS: 21498-08-8 | C11H13Cl3N2O | 295.588 g/mol

Alpha-2 adrenergic agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21498-08-8
Molecular Formula C11H13Cl3N2O
Molecular Weight (g/mol) 295.588
MDL Number MFCD00917022
InChI Key DWWHMKBNNNZGHF-UHFFFAOYSA-N
Synonym lofexidine hydrochloride, lofexidine hcl, lofetensin, loxacor, lofexidine.hcl, loxacor hydrochloride, lofetensin hydrochloride, 2-1-2,6-dichlorophenoxy ethyl-4,5-dihydro-1h-imidazole hydrochloride, lofexidine hydrochloride usan
PubChem CID 30667
IUPAC Name 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole;hydrochloride
SMILES CC(C1=NCCN1)OC2=C(C=CC=C2Cl)Cl.Cl

Specifications

Melting Point 225°C to 232°C
Color Yellow
Quantity 1 g
UN Number UN2811
Merck Index 14,5558
Formula Weight 295.6
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Lofexidine hydrochloride
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only