missing translation for 'onlineSavingsMsg'
Learn More

Locust (Carob) Bean Gum, FCC, Spectrum™ Chemical

Catalog No. 18603963
Change view
Click to view available options
Quantity:
250 g
500 g
2.5 kg
50 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603963 250 g Poly Pail
18603960 500 g Poly Pail
18603961 2.5 kg Poly Pail
18603962 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603963 Supplier Spectrum Chemical Mfg Cor Supplier No. L1135250GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L1135, 9000-40-2

Spectrum™ Chemical Locust (Carob) Bean Gum, FCC is a gelling and thickening agent used in certain foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-40-2

Specifications

CAS Max % 100%
Loss on Drying 14%
Packaging Poly Pail
Quantity 250 g
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Locust (Carob) Bean Gum