Learn More
Locust (Carob) Bean Gum, FCC, Spectrum™ Chemical
L1135, 9000-40-2
Supplier:  Spectrum Chemical Mfg Cor L1135500GM
 View more versions of this product
Catalog No. 18603960
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Locust (Carob) Bean Gum, FCC is a gelling and thickening agent used in certain foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
9000-40-2
14%
500 g
100%
Poly Bottle
FCC
Chemical Identifiers
9000-40-2
Product Suggestions
Product Suggestions