missing translation for 'onlineSavingsMsg'
Learn More

Locostatin

Catalog No. AAJ64425MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64425MF 50 mg
AAJ64425MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64425MF Supplier Thermo Scientific Chemicals Supplier No. J64425MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 133812-16-5 | C14H15NO3 | 245.278 g/mol

Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Locostatin is a cell-permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration

Chemical Identifiers

CAS 133812-16-5
Molecular Formula C14H15NO3
Molecular Weight (g/mol) 245.278
MDL Number MFCD00278769
InChI Key UTZAFVPPWUIPBH-QSLRECBCSA-N
Synonym locostatin, s-4-benzyl-3-but-2-enoyl oxazolidin-2-one, s,e-4-benzyl-3-but-2-enoyl oxazolidin-2-one, 4s-n-crotonyl-4-benzyl-2-oxazolidinone, 4s-3-e-but-2-enoyl-4-benzyl-2-oxazolidinone, 4s-4-benzyl-3-e-but-2-enoyl-1,3-oxazolidin-2-one, cell sheet migration inhibitor, locostatin, 4s-4-benzyl-3-2e-but-2-enoyl-1,3-oxazolidin-2-one, locostatin hplc, 3-crotonyl-4beta-benzyloxazolidine-2-one
PubChem CID 5702600
IUPAC Name (4S)-4-benzyl-3-[(E)-but-2-enoyl]-1,3-oxazolidin-2-one
SMILES CC=CC(=O)N1C(COC1=O)CC2=CC=CC=C2

Specifications

Color White
Quantity 50 mg
Formula Weight 245.27
Physical Form Powder
Chemical Name or Material Locostatin
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only