Learn More
Brady™ Lockout Procedure Station with Binder
Catalog No. 296362252
Change view
Click to view available options
Color:
Red
Catalog No. Color
296362252 Red
1 options
Catalog No. 296362252 Supplier Brady™ Supplier No. 99289
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Lockout Station is perfect for ensuring that all of lockout procedures are readily available to employees. The procedure station's silk-screened backboard is made from durable polystyrene and includes pre-drilled holes and hardware for wall mounting.
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Product Type Procedure Station
Color Red
Material Polystyrene
Includes Backboard with white PVC coated steel binder rack and hardware and a Lockout Procedure Binder (LOSB1). (Procedure Forms sold separately)
Quantity 1 Kit