missing translation for 'onlineSavingsMsg'
Learn More

Poltex™ Locking Medication Box

Catalog No. 22000823
Encompass
Change view
Click to view available options
Dimensions (L x W x H):
5 x 8 x 4 in.
6 x 12 x 6 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
22000823 5 x 8 x 4 in.
22000813 6 x 12 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000823 Supplier Poltex™ Supplier No. DRUGBX854
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions

missing translation for 'validMainframeMsg'

Locking Medication Box is made of super durable 0.25 in. thick PETG construction.

  • Digital combination lock (4-digit code)
  • Mounts under cabinet, to wall, or to countertop
  • Screws and anchors included
  • Choose from two sizes

Specifications

Product Type Storage Case
Color Clear
Material PETG
Dimensions (L x W x H) 5 x 8 x 4 in.