missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Protein LoBind™ Tubes

Catalog No. 13698795
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13698795 2 mL
13698794 1.5 mL
13698793 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13698795 Supplier Eppendorf™ Supplier No. 022431102
Only null left

Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.

  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Snap, screw, and SnapTec™ cap options available.

Specifications

Certifications/Compliance DNA, DNase, RNase, PCR Inhibitors free
Closure Material HDPE
Color Clear
Capacity (Metric) 2 mL
Material Polypropylene
For Use With Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
Includes Safe-lock lid
No. per Pack 100 Pk.
Packaging 100 Tubes (2 Bags x 50 Tubes)
Sterility Non-sterile
RNase Free Yes
Dnase Free Yes
Autoclavable Autoclavable
Shape Cylindrical
Capacity (English) 0.07 oz.
Clean Claims Free of surface coating (e.g. silicone), plasticizers, biocides or mold release agents
Bottom Shape Conical
Closure Type Snap Cap
Product Type Microcentrifuge Tube (PCR Clean, Protein LoBind)
Max. RCF 18,000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Show More Show Less