Learn More
Fisherbrand™ LN2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp LN2 Backup Systems for Ultra-Low Temperature Freezers.
Supplier: Fisherbrand™ 332353G07
Description
- Maintains temperature down to -80°C with liquid nitrogen
Specifications
| Factory Installed | |
| -80°C | |
| Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer |