Learn More
Fisherbrand™ LN2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Catalog No. 332353G03
Change view
Click to view available options
For Use With (Equipment):
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
332353G03 Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
332353G07 Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 332353G03 Supplier Fisherbrand™ Supplier No. 332353G03
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp LN2 Backup Systems for Ultra-Low Temperature Freezers.
  • Maintains temperature down to -80°C with liquid nitrogen
Specifications
Description Factory Installed
Type Rack
Temperature Range -80°C
For Use With (Equipment) Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer