missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ Fixable Olive (557) Viability Kit, for 488 nm excitation

Catalog No. L34977
Encompass
Change view
Click to view available options
Quantity:
200 Assays
400 Assays
80 Assays
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L34977 80 Assays
L34978 200 Assays
L34979 400 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L34977 Supplier Invitrogen™ Supplier No. L34977

The LIVE/DEAD Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry.

  • Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry
  • Cells labeled with LIVE/DEAD Fixable viability dyes can be washed, fixed, permeabilized, and stained for intracellular antigens while maintaining distinct live and dead cell populations
  • The narrow and unique emission spectra are ideal for expanding high-parameter flow cytometry experiments
  • Designed to provide a bright signal with minimal overlap into adjacent channels, simplifying spectral compensation
  • Since live and dead cells can be discriminated using a single flow cytometer channel, these viability dyes are an ideal choice for multi-color experiments.
  • Their unique spectral fingerprints are a valuable addition to spectral flow cytometry experiments
  • Dyes have been conveniently packaged in single-use vials to help ensure dye stability and performance over time
  • Dissolve the dye in the included anhydrous DMSO and use immediately, or store at -20°C, avoiding freeze-thaw cycles
  • The LIVE/DEAD Fixable Dead Cell Stain Sampler Kit provides one vial (40 assays) of each of eight available dead cell stains

Specifications

Quantity 80 Assays
Description LIVE/DEAD™ Fixable Olive (557) Viability Kit, for 488 nm excitation
Format Tubes
Color Olive
Content And Storage Contains 2 vials of LIVE/DEAD fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Compatible Cells Eukaryotic Cells
Excitation/Emission 479/557 nm
Flow Cytometer Laser Lines 488 nm
Solubility DMSO (Dimethylsulfoxide)
Shipping Condition Room Temperature
Product Line LIVE/DEAD
For Use With (Equipment) Flow Cytometer
Detection Method Fluorescence
Conjugate LIVE/DEAD Fixable Olive (557) Dead Cell Stain
For Use With (Application) Flow Cytometry
Product Type Cell Viability Kit
Show More Show Less
What is the concentration of the LIVE/DEAD Fixable dyes?

The concentration of LIVE/DEAD Fixable dyes is proprietary.