missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Live Cell Imaging Solution
GREENER_CHOICE

Catalog No. A59688DJ
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A59688DJ 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A59688DJ Supplier Invitrogen™ Supplier No. A59688DJ
  • ONLINE EXCLUSIVE PRICE
    $49.27
    $40.40 / Each

    Save $8.87

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Live Cell Imaging Solution from Molecular Probes™ is a physiological medium developed for live cell imaging applications that has universal utility in cell-based research—for live cell imaging, dye loading, and wash steps.

The Live Cell Imaging Solution from Molecular Probes™ is a physiological medium developed for live cell imaging applications that has universal utility in cell-based research—for live cell imaging, dye loading, and wash steps. It is an optically clear, physiological solution buffered with HEPES at pH 7.4 that keeps cells healthy for up to 4 hours at ambient atmosphere and temperature. Compared to standard media, Live Cell Imaging Solution provides better clarity, signal-to-noise ratio, and cell viability.

Outstanding Features of Live Cell Imaging Solution:
• Physiological and chemical optimization—providing the ideal solution for live cell fluorescence imaging applications
• Optical clarity—affording more efficient imaging and minimal background
• Extended buffering capacity compared to media—for improved cell viability

Obtain Great Results with a Medium Designed to Meet the Demands of Live Cell Imaging
Live cell imaging can be challenging. Not only must staining be optimized for the particular assay readout, spectral compatibility, and signal-to-noise level, but live cell imaging of dynamic processes and cell physiology requires live observation over time. The choice of medium is particularly important for time-lapse imaging and experiments where cells are exposed to ambient conditions for longer periods. For reliable results, it is essential that cells be healthy and maintained as closely as possible to physiological temperature, pH, oxygen tension, and other conditions. If cells appear unhealthy they should not be used for imaging. Live Cell Imaging Solution has been developed specifically for live cell imaging applications; as a result, it provides image clarity (Fig 1), signal-to-noise ratio (Fig 2), and cell viability that are significantly enhanced compared to standard media. In addition to live cell imaging, the physiological composition of the Live Cell Imaging Solution makes it ideal for other cell-based applications, such as wash steps and dye loading.

Order Info

Shipping Condition: Room temperature

Specifications

Product Line Molecular Probes
Reagent Type Physiological Imaging Buffer
Content And Storage

Store at ≤25°C.

Quantity 500 mL
Shipping Condition Room Temperature
What is the formulation of the Live Cell Imaging Solution (Cat. No. A59688DJ)?

The formulation of the Live Cell Imaging Solution (Cat. No. A59688DJ) is shown below:
140 mM NaCl
2.5 mM KCl
1.8 mM CaCl2
1.0 mM MgCl2
20 mM HEPES
pH 7.4
mOsm = 300

For Research Use Only.