missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Litmus, pure, indicator grade

CAS: 1393-92-6

$158.42 - $507.12

Chemical Identifiers

CAS 1393-92-6
MDL Number MFCD00131528
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC190220250
SDS View Documents
Thermo Scientific Chemicals
190220250
25 g Glass bottle
Each for $158.42
Only null left
 
AC190221000
SDS View Documents
Thermo Scientific Chemicals
190221000
100 g Glass bottle
Each for $507.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

1393-92-6
MFCD00131528

Specifications

1393-92-6
15, 5603
Authentic
Glass bottle
Black/Blue to Blue
Powder or Granules
MFCD00131528
Solubility in water: soluble. Other solubilities: partially soluble in alcohol
Pure
from pH 4.8 (red) to pH 8.3 (blue)
25 g
Litmus
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only