missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Litmus, pure, indicator grade

Catalog No. AC190225000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190225000 500 g Glass bottle
AC190220250 25 g Glass bottle
AC190221000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC190225000 Supplier Thermo Scientific Chemicals Supplier No. 190225000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1393-92-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 1393-92-6
MDL Number MFCD00131528

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Visual Transition Interval from pH 4.8 (red) to pH 8.3 (blue)
Merck Index 15, 5603
Color Black/Blue to Blue
Solubility Information Solubility in water: soluble. Other solubilities: partially soluble in alcohol
Quantity 500 g
Physical Form Powder or Granules
Grade Pure
Chemical Name or Material Litmus

RUO – Research Use Only