missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Litmus Paper
Litmus Paper available as vial of 100.
$7.07 - $7.13
Specifications
| For Use With (Application) | pH Indicator |
|---|---|
| Quantity | 100 Pk. |
| Type | pH Papers |
Description
- pH indicator
- Available as vial of 100
Specifications
| pH Indicator | |
| 100 Pk. | |
| pH Papers |