missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Litmus Paper

Catalog No. S24483
Change view
Click to view available options
Color:
Blue
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S24483 Red
S24482 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S24483 Supplier Eisco™ Supplier No. LITMUSRED
Edge

Litmus Paper available as vial of 100.

  • pH indicator
  • Available as vial of 100

Specifications

Color Red
pH Range 6.8 to 8.1
For Use With (Application) pH Indicator
Quantity 100 Pk.
Type pH Papers