missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Litmus Paper

Catalog No. S24482
Change view
Click to view available options
Color:
Blue
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S24482 Blue
S24483 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S24482 Supplier Eisco™ Supplier No. LITMUSBLUE

Litmus Paper available as vial of 100.

  • pH indicator
  • Available as vial of 100

Specifications

Color Blue
pH Range 4.3 to 6.8
For Use With (Application) pH Indicator
Quantity 100 Pk.
Type pH Papers