missing translation for 'onlineSavingsMsg'
Learn More

Lithium trifluoroacetate, 97%

Catalog No. p-7046917
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1680109 10 g
AAL1680118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1680109 Supplier Thermo Scientific Chemicals Supplier No. L1680109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2923-17-3 | C2F3LiO2 | 119.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2923-17-3
Molecular Formula C2F3LiO2
Molecular Weight (g/mol) 119.96
MDL Number MFCD00013216,MFCD00149663
InChI Key HSFDLPWPRRSVSM-UHFFFAOYSA-M
Synonym lithium trifluoroacetate, acetic acid, trifluoro-, lithium salt, lithium 2,2,2-trifluoroacetate, lithiumtrifluoroacetate, acmc-1cfal, lithotab trifluoroacetate, acetic acid, 2,2,2-trifluoro-, lithium salt 1:1, lithium trifluoroacetate, monohydrate, lithium 1+ trifluoroacetate, lithio 2,2,2-trifluoroacetate
PubChem CID 23661853
IUPAC Name lithium;2,2,2-trifluoroacetate
SMILES [Li+].[O-]C(=O)C(F)(F)F

Specifications

Quantity 10 g
UN Number UN2811
Formula Weight 119.96
Percent Purity 97%
Chemical Name or Material Lithium trifluoroacetate
Hazard Category H300
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 220-878-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only