missing translation for 'onlineSavingsMsg'
Learn More

Lithium tri-sec-butylborohydride, 1.0M solution in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA43034AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43034AE 100 mL
AA43034AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43034AE Supplier Thermo Scientific Chemicals Supplier No. 043034AE
Only null left

As a reducing agent of olefinic groups and ketones | CAS: 38721-52-7 | C12H28BLi | 190.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 38721-52-7
Molecular Formula C12H28BLi
Molecular Weight (g/mol) 190.11
MDL Number MFCD00011708
InChI Key ACJKNTZKEFMEAK-UHFFFAOYNA-N
Synonym lithium tri-sec-butylhydroborate, l-selectridesolution, lithium tri-sec-butylborohydride solution, l-selectride™ solution, lithium tri butan-2-yl boron 1-, lithium tri-sec-butylborohydride l-selectride , 1m in thf, lithium tri-sec-butylborohydride 1.0 m solution in thf, spcseal, lithium tri-sec-butylborohydride, 1.0m solution in thf, packaged under argon in resealable chemseal bottles
IUPAC Name lithium(1+) tris(butan-2-yl)boranuide
SMILES [Li+].CCC(C)[BH-](C(C)CC)C(C)CC

Specifications

Color Colorless to Yellow
Density 0.870 g/mL
Flash Point −17°C (1°F)
Packaging ChemSeal™ bottles
Linear Formula LiB[CH(CH3)CH2CH3]3H
Quantity 100 mL
UN Number UN3394
Beilstein 4007327
Sensitivity Moisture sensitive
Merck Index 14,8427
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 190.11
Concentration or Composition (by Analyte or Components) 1.0M solution in THF
Physical Form Liquid
Chemical Name or Material Lithium tri-sec-butylborohydride
Show More Show Less
Hazard Category H225-H260-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H250-H260-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: I
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only