missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct

Catalog No. AA3544502
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3544502 0.5 g
AA3544504 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3544502 Supplier Thermo Scientific Chemicals Supplier No. 03544502

CAS: 75965-35-4 | C36H50BLiO6 | 596.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75965-35-4
Molecular Formula C36H50BLiO6
Molecular Weight (g/mol) 596.54
MDL Number MFCD00013311
InChI Key ADYUZFWVPWDPFK-UHFFFAOYSA-N
Synonym lithium tetraphenylborate tris 1,2-dimethoxyethane, tetraphenylboron lithium tris 1,2-dimethoxyethane, lithium tetraphenylborate tris 1,2-dimethoxyethane adduct, lithium tetraphenylborate 1--1,2-dimethoxyethane 1/1/3, lithium 1+ tris 1,2-dimethoxyethane tetraphenylborate
PubChem CID 23681136
IUPAC Name lithium;1,2-dimethoxyethane;tetraphenylboranuide
SMILES [Li+].[B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.COCCOC.COCCOC.COCCOC

Specifications

Color White
Linear Formula (C6H5)4BLi·3CH3OCH2CH2OCH3
Quantity 0.5 g
UN Number UN1325
Sensitivity Moisture sensitive
Formula Weight 595.54 (326.18 Anhydrous)
Physical Form Powder
Chemical Name or Material Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only