Learn More
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
CAS: 371162-53-7 | 2·5 C4H10O | 871.28 g/mol
Supplier: Thermo Scientific Chemicals 458530010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Lithium tetrakis(pentafluorophenyl)borate ethyl etherate | |
| 371162-53-7 | |
| 2·5 C4H10O | |
| KPLZKJQZPFREPG-UHFFFAOYSA-N | |
| lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide | |
| 23701484 |
| 117.0°C to 122.0°C | |
| 1 g | |
| lithium tetrakis pentafluorophenyl borate ethyl etherate, lithium tetrakis pentafluorophenyl borate, tetrakis pentafluorophenyl boron lithium ethyl etherate, lithium tetrakis pentafluorophenyl borate-ethyl ether complex, lithium tetrakis pentafluorophenyl borate ethyl etherate 2:2:5, lithium tetrakis pentafluorophenyl borate diethylether complex 1:2.5 | |
| [Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC | |
| 871.28 | |
| 871.28 |
Chemical Identifiers
| 371162-53-7 | |
| 871.28 | |
| lithium tetrakis pentafluorophenyl borate ethyl etherate, lithium tetrakis pentafluorophenyl borate, tetrakis pentafluorophenyl boron lithium ethyl etherate, lithium tetrakis pentafluorophenyl borate-ethyl ether complex, lithium tetrakis pentafluorophenyl borate ethyl etherate 2:2:5, lithium tetrakis pentafluorophenyl borate diethylether complex 1:2.5 | |
| lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide |
| 2·5 C4H10O | |
| KPLZKJQZPFREPG-UHFFFAOYSA-N | |
| 23701484 | |
| [Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
RUO – Research Use Only