missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetrafluoroborate, 98%, pure

Catalog No. AC223770100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223770100 10 g Plastic bottle
AC223770500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223770100 Supplier Thermo Scientific Chemicals Supplier No. 223770100

CAS: 14283-07-9 | BF4Li | 93.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14283-07-9
Molecular Formula BF4Li
Molecular Weight (g/mol) 93.74
MDL Number MFCD00011087
InChI Key UFXJWFBILHTTET-UHFFFAOYSA-N
Synonym lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n
PubChem CID 4298216
IUPAC Name lithium(1+) tetrafluoroboranuide
SMILES [Li+].F[B-](F)(F)F

Specifications

Melting Point 275°C
Color Gray to White
Physical Form Crystalline Powder and/or Crystals
Quantity 10 g
Assay Percent Range 98%
Linear Formula LiBF4
Fieser 05,416; 10,248
Merck Index 15, 5599
Solubility Information Solubility in water: soluble
Formula Weight 93.75
Percent Purity 98%
Grade Pure
Packaging Plastic bottle
Specific Gravity 0.852
Density 0.8520g/mL
Chemical Name or Material Lithium tetrafluoroborate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-178-9
TSCA TSCA

RUO – Research Use Only