missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetrachloropalladate(II)

Catalog No. p-7049648
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1731603 1 g
AA1731606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1731603 Supplier Thermo Scientific Chemicals Supplier No. 01731603
Only null left

CAS: 15525-45-8

It act as a catalyst in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15525-45-8
MDL Number MFCD00011082

Specifications

Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 262.09
Sensitivity Moisture sensitive
Chemical Name or Material Lithium tetrachloropalladate(II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-567-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only