missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetraborate trihydrate, 96%

Catalog No. AAA1035836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1035836 500 g
AAA1035822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1035836 Supplier Thermo Scientific Chemicals Supplier No. A1035836

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37195-62-3 | Li2B4O7·3H2O

It is employed in the conductances of some potassium and sodium salts in dimethylformamide at 25°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the conductances of some potassium and sodium salts in dimethylformamide at 25°C.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 37195-62-3
Molecular Formula Li2B4O7·3H2O
MDL Number MFCD00167253
Synonym lithium borate trihydrate, acmc-1ajhy, trilithium 1+ trihydrate borate, trilithium 1+ ion trihydrate borate

Specifications

Quantity 500 g
Merck Index 14,5524
Solubility Information Soluble in water.
Formula Weight 223.17 (169.12 Anhydrous)
Percent Purity 96%
Chemical Name or Material Lithium tetraborate trihydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-514-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only