missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetraborate, 99% (metals basis)

CAS: 12007-60-2 | B4Li2O7 | 169.11 g/mol

$85.45 - $967.38

Chemical Identifiers

CAS 12007-60-2
Molecular Formula B4Li2O7
Molecular Weight (g/mol) 169.11
MDL Number MFCD00011083
InChI Key RDZCISUGRQUANA-UHFFFAOYSA-N
IUPAC Name dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1341918
View Documents
Thermo Scientific Chemicals
01341918
50 g
Each for $85.45
Only null left
 
AA1341930
View Documents
Thermo Scientific Chemicals
01341930
250 g
Each for $291.27
Only null left
 
AA13419A1
View Documents
Thermo Scientific Chemicals
013419A1
1 kg
Each for $967.38
Only null left
 
Description

Description

Used in the laboratory as LB (Lithium borate) buffer for gel electrophoresis of DNA and RNA. Employed in the borax fusion method to modify mineral powder samples for analysis by WDXRF (Wavelength Dispersive X-Ray Fluorescence) Spectroscopy. Acts as an ingredient in the manufacture of glasses and ceramics. Useful fod dissolving basic oxides such as CaO and MgO. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12007-60-2
169.11
RDZCISUGRQUANA-UHFFFAOYSA-N
[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
B4Li2O7
MFCD00011083
dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)

Specifications

930°C
12007-60-2
50 g
B4Li2O7
Moderately soluble in water.
[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
169.11
2.4 g/cm3 at 20°C
White
Powder
99% (Metals basis)
MFCD00011083
RDZCISUGRQUANA-UHFFFAOYSA-N
dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
169.12
Lithium tetraborate
Videos
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 234-514-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only