missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium tetraborate, 98+%, for analysis
CAS: 12007-60-2 | B4Li2O7 | 169.11 g/mol
$316.94 - $946.18
Chemical Identifiers
| CAS | 12007-60-2 |
|---|---|
| Molecular Formula | B4Li2O7 |
| Molecular Weight (g/mol) | 169.11 |
| MDL Number | MFCD00011083 |
| InChI Key | RDZCISUGRQUANA-UHFFFAOYSA-N |
| IUPAC Name | dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
| SMILES | [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC206942500
|
Thermo Scientific Chemicals
206942500 |
250 g | Plastic bottle |
Each for $316.94
|
|
||||
|
AC206940010
|
Thermo Scientific Chemicals
206940010 |
1 kg | Plastic bottle |
Each for $946.18
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12007-60-2 | |
| 169.11 | |
| RDZCISUGRQUANA-UHFFFAOYSA-N | |
| [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
| B4Li2O7 | |
| MFCD00011083 | |
| dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
Specifications
| 930.0°C | |
| White | |
| 250 g | |
| B4Li2O7 | |
| MFCD00011083 | |
| Solubility in water: 28g/L (20°C). Other solubilities: 52g/L (100°C) | |
| [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 | |
| 169.11 | |
| ≥98% | |
| 0.5% max. (200°C) | |
| Lithium tetraborate, p.a. |
| 12007-60-2 | |
| Powder | |
| 98+% | |
| Li2B4O7 | |
| 15, 5580 | |
| RDZCISUGRQUANA-UHFFFAOYSA-N | |
| dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) | |
| 169.12 | |
| Analytical | |
| Plastic bottle |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 234-514-3
TSCA : TSCA
RUO – Research Use Only