missing translation for 'onlineSavingsMsg'
Learn More

Lithium tert-butoxide, 99%, Thermo Scientific™

Catalog No. AC301225000
Change view
Click to view available options
500g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC301225000 500g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC301225000 Supplier Thermo Scientific Chemicals Supplier No. 301225000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 1907-33-1
Density 0.8900g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic Bottle
Molecular Formula C4H9LiO
Linear Formula LiOCH(CH3)3
Quantity 500g
UN Number 2925
Specific Gravity 0.89
Synonym lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
Sublimation Point 283 °C
Solubility Information Solubility in water: vigorous reaction
InChI Key LZWQNOHZMQIFBX-UHFFFAOYSA-N
SMILES [Li+].CC(C)(C)[O-]
IUPAC Name lithium;2-methylpropan-2-olate
Molecular Weight (g/mol) 80.06
PubChem CID 23664764
Formula Weight 80.06
Percent Purity 99%
Chemical Name or Material Lithium tert-butoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Harmful if swallowed.
Reacts violently with water.
Precautionary Statement GHS P Statement
Keep cool.
Protect from sunlight.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if
EINECSNumber 217-611-5

RUO – Research Use Only